Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chaf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013733.3 | NP_038761.1 | 10 - 11 | 1885 | 91 | |
GenBank mRNA | AJ132771.2 | - | 10 - 11 | 1789 | 91 | |
AK034839.1 | - | 10 - 11 | 1886 | 91 | ||
AK132551.1 | - | 1709 | 91 | |||
AK161016.1 | - | 713 | 91 | |||
BC053740.1 | - | 10 - 11 | 1856 | 6504838 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |