Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172619.3 | NP_766207.2 | 7 - 8 | 1204 | 81 | |
NR_037707.1 | 7 - 8 | 1209 | 81 | |||
NR_037708.1 | 7 - 8 | 1209 | 81 | |||
GenBank mRNA | AK042525.1 | - | 7 - 8 | 1197 | 81 | |
AK048600.1 | - | 3 - 4 | 398 | 81 | ||
AK076334.1 | - | 7 - 8 | 1184 | 81 | ||
BC056427.1 | - | 7 - 8 | 1179 | 81 | ||
BC082773.1 | - | 7 - 8 | 1201 | 81 | ||
BC145353.1 | - | 6 - 7 | 1116 | 81 | ||
BC145355.1 | - | 6 - 7 | 1116 | 81 | ||
BQ571453.1 | - | 1 - 2 | 140 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |