Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009059.2 | NP_033085.2 | 9 - 10 | 1403 | 123 | |
GenBank mRNA | AK144440.1 | - | 9 - 10 | 1405 | 123 | |
BC068121.1 | - | 9 - 10 | 1403 | 6415928 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |