Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700005711.mm.1 |
NM_001205214.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005711.mm.1 |
NM_001205214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005711.mm.1 |
NM_001205215.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005711.mm.1 |
NM_001205215.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005711.mm.1 |
NM_001205216.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005711.mm.1 |
NM_001205216.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005711.mm.1 |
NM_011306.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005711.mm.1 |
NM_011306.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005711.mm.1 |
TC1700000611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005713.mm.1 |
NM_001205214.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005713.mm.1 |
NM_001205214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005713.mm.1 |
NM_001205215.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005713.mm.1 |
NM_001205215.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005713.mm.1 |
NM_001205216.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005713.mm.1 |
NM_001205216.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005713.mm.1 |
NM_011306.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005713.mm.1 |
NM_011306.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005713.mm.1 |
TC1700000611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005716.mm.1 |
NM_001205214.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005716.mm.1 |
NM_001205214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005716.mm.1 |
NM_001205215.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005716.mm.1 |
NM_001205215.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005716.mm.1 |
NM_001205216.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005716.mm.1 |
NM_001205216.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005716.mm.1 |
NM_011306.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005716.mm.1 |
NM_011306.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005716.mm.1 |
TC1700000611.mm.1 |
MTA TC |