Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161730.1 | NP_001155202.1 | 10 - 11 | 2066 | 82 | |
NM_013683.2 | NP_038711.2 | 9 - 10 | 2150 | 82 | ||
GenBank mRNA | AK040919.1 | - | 7 - 8 | 3113 | 82 | |
AK151481.1 | - | 9 - 10 | 1905 | 82 | ||
AK152953.1 | - | 9 - 10 | 1850 | 82 | ||
AK153890.1 | - | 9 - 10 | 2154 | 82 | ||
AK162731.1 | - | 9 - 10 | 1950 | 82 | ||
AK166046.1 | - | 9 - 10 | 2152 | 82 | ||
AK167485.1 | - | 9 - 10 | 2152 | 82 | ||
AK171363.1 | - | 10 - 11 | 1821 | 82 | ||
BC013802.1 | - | 9 - 10 | 1561 | 3257566 | 82 | |
BC024897.1 | - | 9 - 10 | 1673 | 3257734 | 82 | |
BC031584.1 | - | 5 - 6 | 2285 | 82 | ||
KJ650227.1 | - | 9 - 10 | 1826 | 82 | ||
M55637.1 | - | 9 - 10 | 1542 | 82 | ||
U60018.1 | - | 9 - 10 | 1826 | 82 | ||
U60019.1 | - | 9 - 10 | 1826 | 82 | ||
U60020.1 | - | 9 - 10 | 1826 | 82 | ||
U60021.1 | - | 9 - 10 | 1826 | 82 | ||
U60022.1 | - | 9 - 10 | 1826 | 82 | ||
U60023.1 | - | 9 - 10 | 1826 | 82 | ||
X59615.1 | - | 9 - 10 | 1385 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |