Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053194.4 | NP_444424.1 | 1 - 2 | 161 | 58 | |
GenBank mRNA | AF288813.1 | - | 1 - 2 | 160 | 58 | |
AK031466.1 | - | 1 - 2 | 108 | 58 | ||
AK080192.1 | - | 1 - 2 | 104 | 58 | ||
AK155282.1 | - | 1 - 2 | 136 | 58 | ||
AK165536.1 | - | 1 - 2 | 138 | 58 | ||
AK167746.1 | - | 1 - 2 | 164 | 58 | ||
AK169367.1 | - | 1 - 2 | 125 | 58 | ||
BC022917.1 | - | 1 - 2 | 138 | 4460521 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |