Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brsk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009929.3 | NP_001009929.1 | 7 - 8 | 976 | 65 | |
NM_001009930.3 | NP_001009930.1 | 7 - 8 | 976 | 65 | ||
NM_001276763.1 | NP_001263692.1 | 7 - 8 | 976 | 65 | ||
NM_029426.2 | NP_083702.1 | 7 - 8 | 976 | 65 | ||
GenBank mRNA | AK158574.1 | - | 7 - 8 | 725 | 65 | |
AK173268.1 | - | 6 - 7 | 551 | 65 | ||
AY533672.1 | - | 7 - 8 | 641 | 65 | ||
AY533673.1 | - | 7 - 8 | 641 | 65 | ||
AY533674.1 | - | 7 - 8 | 641 | 65 | ||
AY660739.1 | - | 7 - 8 | 976 | 65 | ||
BC056498.1 | - | 4 - 5 | 252 | 65 | ||
JF707771.1 | - | 7 - 8 | 641 | 65 | ||
JF707772.1 | - | 7 - 8 | 641 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |