Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110794.1 | NP_001104264.1 | 9 - 10 | 1147 | 85 | |
NM_009674.3 | NP_033804.2 | 9 - 10 | 1262 | 85 | ||
GenBank mRNA | AK032013.1 | - | 9 - 10 | 1038 | 85 | |
AK134589.1 | - | 10 - 11 | 1032 | 85 | ||
AK149654.1 | - | 9 - 10 | 954 | 85 | ||
AK151438.1 | - | 9 - 10 | 956 | 85 | ||
AK161613.1 | - | 9 - 10 | 956 | 85 | ||
AK167588.1 | - | 9 - 10 | 930 | 85 | ||
AK169199.1 | - | 9 - 10 | 929 | 85 | ||
BC008997.1 | - | 9 - 10 | 946 | 3157926 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |