Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bahd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045523.1 | NP_001038988.1 | 4 - 5 | 2105 | 62 | |
GenBank mRNA | AK129243.1 | - | 4 - 5 | 2263 | 62 | |
AK152597.1 | - | 4 - 5 | 2044 | 62 | ||
BC047433.1 | - | 3 - 4 | 854 | 62 | ||
BC100358.1 | - | 4 - 5 | 2267 | 30848279 | 62 | |
BC141046.1 | - | 3 - 4 | 1968 | 9055658 | 62 | |
BC145233.1 | - | 3 - 4 | 1968 | 9053785 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |