Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183282.2 | NP_899105.2 | 1 | 1327 | 73 | |
GenBank mRNA | AK006498.1 | - | 1 | 1320 | 73 | |
BC089487.1 | - | 1 | 831 | 73 | ||
BC100422.1 | - | 1 | 646 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |