Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-T24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008207.3 | NP_032233.2 | 1 - 2 | 105 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |