Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2056973 |
NM_001033405.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P107483 |
NM_001033405.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P107483 |
AK037593.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P107483 |
DQ087184.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P107483 |
DQ341272.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
546241 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
478338 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
53572 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
895544 |
AK015686.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
895544 |
NM_029131.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
788232 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
183396 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
198523 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
228737 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
734395 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
40567 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2056973 |
NM_001033405.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P107483 |
NM_001033405.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P107483 |
AK037593.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P107483 |
DQ087184.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P107483 |
DQ341272.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216713 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216734 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216722 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216729 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216720 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216727 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216715 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216731 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216736 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216702 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216796 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216721 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216707 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216703 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216710 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216735 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216737 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216708 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216733 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216716 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216728 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216704 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216718 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216725 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216724 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216726 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31216738 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08794511 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03327837 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03327851 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P03327851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216694 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216777 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216727 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216799 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216849 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216758 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216853 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216734 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216837 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216783 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216855 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216720 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216781 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216750 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216766 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216840 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216859 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216753 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216772 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216770 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216701 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216845 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216713 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216763 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216722 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216857 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216865 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216755 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216795 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216775 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216738 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216768 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216862 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216785 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216733 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216792 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216867 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216788 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216868 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216866 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216804 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216864 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31216724 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216740 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216710 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216756 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216763 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13472436 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13372812 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216750 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216717 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216804 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13345765 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216704 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216755 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216731 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216760 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216728 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216797 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13059604 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13472435 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216754 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13345559 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216787 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13381055 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216726 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13529651 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216701 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216716 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216702 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216738 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216781 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216753 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216741 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13529650 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216791 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216724 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13182911 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216762 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216802 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216736 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216743 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216715 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216705 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216784 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13294605 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216727 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216707 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13472439 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216703 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216733 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13182903 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13529652 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13529874 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216758 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216713 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13504890 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216725 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216792 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216800 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216788 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216699 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216799 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216734 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216776 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13182906 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31216700 |
- |
- |