Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dfnb59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080711.2 | NP_001074180.1 | 2 - 3 | 550 | 72 | |
GenBank mRNA | DQ365828.1 | - | 2 - 3 | 550 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Mus musculus Probe spans exons |