Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285507.1 | NP_001272436.1 | 1 - 2 | 143 | 71 | |
NM_001285512.1 | NP_001272441.1 | 1 - 2 | 143 | 71 | ||
NM_001285513.1 | NP_001272442.1 | 1 - 2 | 143 | 71 | ||
NM_009939.3 | NP_034069.2 | 1 - 2 | 143 | 71 | ||
GenBank mRNA | AF071312.1 | - | 1 - 2 | 53 | 71 | |
AF087688.1 | - | 1 - 2 | 50 | 71 | ||
AK010383.1 | - | 1 - 2 | 69 | 71 | ||
AK037757.1 | - | 1 - 2 | 69 | 71 | ||
AK084421.1 | - | 1 - 2 | 64 | 71 | ||
AK132207.1 | - | 1 - 2 | 60 | 71 | ||
AK132776.1 | - | 1 - 2 | 80 | 71 | ||
AK145225.1 | - | 1 - 2 | 54 | 71 | ||
AK146153.1 | - | 1 - 2 | 54 | 71 | ||
BC023096.1 | - | 1 - 2 | 48 | 3675508 | 71 | |
CJ052709.1 | - | 1 - 2 | 143 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |