Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK144560.1 | - | 1 - 2 | 376 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |