Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111142.1 | NP_001104612.1 | 2 - 3 | 609 | 69 | |
GenBank mRNA | BC147714.1 | - | 2 - 3 | 584 | 9087473 | 69 |
BC172136.1 | - | 2 - 3 | 578 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |