Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009085.2 | NP_033111.2 | 1 - 2 | 156 | 64 | |
GenBank mRNA | AK132114.1 | - | 1 - 2 | 98 | 64 | |
AK146240.1 | - | 1 - 2 | 106 | 64 | ||
BC130018.1 | - | 1 - 2 | 157 | 40140532 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |