Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291012.1 | NP_001277941.1 | 2 - 3 | 176 | 120 | |
NM_001291013.1 | NP_001277942.1 | 2 - 3 | 176 | 120 | ||
NM_016902.4 | NP_058598.1 | 2 - 3 | 176 | 120 | ||
GenBank mRNA | AF127180.1 | - | 2 - 3 | 139 | 120 | |
AK159627.1 | - | 2 - 3 | 153 | 120 | ||
BC118953.2 | - | 2 - 3 | 139 | 40093235 | 120 | |
BY147655.1 | - | 2 - 3 | 177 | 120 | ||
EF174553.3 | - | 2 - 3 | 148 | 120 | ||
EF174554.2 | - | 2 - 3 | 148 | 120 | ||
FJ767896.1 | - | 2 - 3 | 139 | 120 | ||
FJ767897.1 | - | 2 - 3 | 139 | 120 | ||
FJ767898.1 | - | 2 - 3 | 139 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |