Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038624.1 | NP_001033713.1 | 2 - 3 | 390 | 108 | |
NM_178780.3 | NP_848895.2 | 2 - 3 | 390 | 108 | ||
GenBank mRNA | AK053760.1 | - | 2 - 3 | 371 | 108 | |
AK082275.1 | - | 2 - 3 | 371 | 108 | ||
AK134663.1 | - | 2 - 3 | 372 | 108 | ||
AK138641.1 | - | 2 - 3 | 389 | 108 | ||
AK149334.1 | - | 2 - 3 | 378 | 108 | ||
BY241896.1 | - | 2 - 3 | 397 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |