Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200006687.mm.1 |
NM_177139.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006687.mm.1 |
NM_177139.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006687.mm.1 |
NR_033304.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006687.mm.1 |
NR_033304.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006687.mm.1 |
TC0200000804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006685.mm.1 |
NM_177139.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006685.mm.1 |
NM_177139.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006685.mm.1 |
NR_033304.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006685.mm.1 |
NR_033304.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006685.mm.1 |
TC0200000804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006689.mm.1 |
NR_033304.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006689.mm.1 |
NR_033304.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006689.mm.1 |
TC0200000804.mm.1 |
MTA TC |