Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700006093.mm.1 |
NM_001111094.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006093.mm.1 |
NM_001111094.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006093.mm.1 |
TC1700000630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006091.mm.1 |
NM_001111094.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006091.mm.1 |
NM_001111094.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006091.mm.1 |
TC1700000630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006094.mm.1 |
NM_001111094.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006094.mm.1 |
NM_001111094.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006094.mm.1 |
TC1700000630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006083.mm.1 |
NM_001111094.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006083.mm.1 |
NM_001111094.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006083.mm.1 |
TC1700000630.mm.1 |
MTA TC |