Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hey1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010423.2 | NP_034553.2 | 2 - 3 | 330 | 55 | |
GenBank mRNA | AB041590.1 | - | 2 - 3 | 223 | 55 | |
AF151521.1 | - | 2 - 3 | 278 | 55 | ||
AF172286.1 | - | 2 - 3 | 311 | 55 | ||
AF176423.1 | - | 2 - 3 | 292 | 55 | ||
AF232241.1 | - | 2 - 3 | 310 | 55 | ||
AJ243895.1 | - | 2 - 3 | 278 | 55 | ||
AK011149.1 | - | 2 - 3 | 332 | 55 | ||
AK140212.1 | - | 2 - 3 | 331 | 55 | ||
BC086635.1 | - | 2 - 3 | 316 | 6809680 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |