Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164190.1 | NP_001157662.1 | 6 - 7 | 636 | 110 | |
NM_001164191.1 | NP_001157663.1 | 6 - 7 | 527 | 110 | ||
NM_001164192.1 | NP_001157664.1 | 6 - 7 | 527 | 110 | ||
NM_019926.3 | NP_064310.2 | 6 - 7 | 516 | 110 | ||
GenBank mRNA | AF073996.1 | - | 6 - 7 | 517 | 110 | |
AK016445.1 | - | 6 - 7 | 637 | 110 | ||
AK031794.1 | - | 6 - 7 | 498 | 110 | ||
AK166452.1 | - | 6 - 7 | 495 | 110 | ||
AK171475.1 | - | 6 - 7 | 505 | 110 | ||
BC051022.1 | - | 6 - 7 | 485 | 110 | ||
BC066847.1 | - | 6 - 7 | 474 | 110 | ||
BC090984.1 | - | 6 - 7 | 494 | 30246206 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |