Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300006739.mm.1 |
NM_172589.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006739.mm.1 |
NM_172589.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006739.mm.1 |
TC1300001056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006736.mm.1 |
NM_172589.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006736.mm.1 |
NM_172589.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006736.mm.1 |
TC1300001056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006737.mm.1 |
NM_172589.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006737.mm.1 |
NM_172589.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006737.mm.1 |
TC1300001056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006745.mm.1 |
NM_172589.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006745.mm.1 |
NM_172589.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006745.mm.1 |
TC1300001056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006746.mm.1 |
NM_172589.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006746.mm.1 |
NM_172589.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006746.mm.1 |
TC1300001056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006747.mm.1 |
NM_172589.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006747.mm.1 |
NM_172589.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006747.mm.1 |
TC1300001056.mm.1 |
MTA TC |