Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146043.2 | NP_001139515.1 | 6 - 7 | 542 | 75 | |
NM_001271463.1 | NP_001258392.1 | 6 - 7 | 542 | 75 | ||
NM_001271464.1 | NP_001258393.1 | 6 - 7 | 542 | 75 | ||
NM_001271465.1 | NP_001258394.1 | 6 - 7 | 569 | 75 | ||
NM_177266.5 | NP_796240.3 | 6 - 7 | 575 | 75 | ||
GenBank mRNA | AK028450.1 | - | 6 - 7 | 528 | 75 | |
AK035819.1 | - | 6 - 7 | 481 | 75 | ||
AK044371.1 | - | 6 - 7 | 543 | 75 | ||
AK083441.1 | - | 6 - 7 | 507 | 75 | ||
AK161797.1 | - | 6 - 7 | 478 | 75 | ||
AK165871.1 | - | 6 - 7 | 536 | 75 | ||
BC138233.1 | - | 6 - 7 | 473 | 8861252 | 75 | |
BC145208.1 | - | 6 - 7 | 473 | 9053759 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |