Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146043.2 | NP_001139515.1 | 9 - 10 | 782 | 63 | |
NM_001271463.1 | NP_001258392.1 | 9 - 10 | 782 | 63 | ||
NM_001271464.1 | NP_001258393.1 | 9 - 10 | 782 | 63 | ||
NM_001271465.1 | NP_001258394.1 | 9 - 10 | 809 | 63 | ||
NM_177266.5 | NP_796240.3 | 9 - 10 | 815 | 63 | ||
GenBank mRNA | AK028450.1 | - | 9 - 10 | 768 | 63 | |
AK035819.1 | - | 9 - 10 | 721 | 63 | ||
AK044371.1 | - | 9 - 10 | 783 | 63 | ||
AK083441.1 | - | 9 - 10 | 747 | 63 | ||
AK161797.1 | - | 9 - 10 | 718 | 63 | ||
AK165871.1 | - | 9 - 10 | 776 | 63 | ||
BC027341.1 | - | 9 - 10 | 766 | 3984129 | 63 | |
BC138233.1 | - | 9 - 10 | 713 | 8861252 | 63 | |
BC145208.1 | - | 9 - 10 | 713 | 9053759 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |