Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162945.1 | NP_001156417.1 | 5 - 6 | 499 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |