Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022011.4 | NP_071294.3 | 9 - 10 | 749 | 137 | |
GenBank mRNA | AB221674.1 | - | 8 - 9 | 563 | 137 | |
AK154739.1 | - | 8 - 9 | 1071 | 137 | ||
BC016231.1 | - | 9 - 10 | 704 | 4488798 | 137 | |
BC053382.1 | - | 9 - 10 | 713 | 30047823 | 137 | |
DV662169.1 | - | 7 - 8 | 489 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |