Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252629.1 | NP_001239558.1 | 5 - 6 | 709 | 58 | |
NM_011420.2 | NP_035550.1 | 5 - 6 | 645 | 58 | ||
GenBank mRNA | AK144778.1 | - | 5 - 6 | 646 | 58 | |
AK152495.1 | - | 5 - 6 | 619 | 58 | ||
AK167832.1 | - | 5 - 6 | 643 | 58 | ||
BC045158.1 | - | 5 - 6 | 609 | 6486685 | 58 | |
EU234021.1 | - | 5 - 6 | 709 | 58 | ||
U63294.1 | - | 5 - 6 | 623 | 58 | ||
U77714.1 | - | 5 - 6 | 606 | 58 | ||
Y12835.1 | - | 5 - 6 | 631 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |