Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mccc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030026.2 | NP_084302.1 | 10 - 11 | 1125 | 67 | |
GenBank mRNA | AK019760.1 | - | 4 - 5 | 306 | 67 | |
AK132265.1 | - | 10 - 11 | 1105 | 67 | ||
AK143233.1 | - | 10 - 11 | 1126 | 67 | ||
AK145564.1 | - | 10 - 11 | 1077 | 67 | ||
AK146844.1 | - | 10 - 11 | 1085 | 67 | ||
AK146865.1 | - | 10 - 11 | 1075 | 67 | ||
BC040098.1 | - | 2 - 3 | 138 | 67 | ||
BC141193.1 | - | 10 - 11 | 1091 | 9055805 | 67 | |
BC141194.1 | - | 10 - 11 | 1091 | 9055806 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |