Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mccc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030026.2 | NP_084302.1 | 11 - 12 | 1193 | 139 | |
GenBank mRNA | AK019760.1 | - | 5 - 6 | 374 | 139 | |
AK132265.1 | - | 11 - 12 | 1173 | 139 | ||
AK143233.1 | - | 11 - 12 | 1194 | 139 | ||
AK145564.1 | - | 11 - 12 | 1145 | 139 | ||
AK146844.1 | - | 11 - 12 | 1153 | 139 | ||
AK146865.1 | - | 11 - 12 | 1143 | 139 | ||
BC040098.1 | - | 3 - 4 | 206 | 138 | ||
BC058987.1 | - | 3 - 4 | 207 | 139 | ||
BC141193.1 | - | 11 - 12 | 1159 | 9055805 | 139 | |
BC141194.1 | - | 11 - 12 | 1159 | 9055806 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |