Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mccc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030026.2 | NP_084302.1 | 15 - 16 | 1611 | 98 | |
GenBank mRNA | AK132265.1 | - | 15 - 16 | 1591 | 98 | |
AK143233.1 | - | 15 - 16 | 1612 | 98 | ||
AK145564.1 | - | 15 - 16 | 1563 | 98 | ||
AK146844.1 | - | 15 - 16 | 1571 | 98 | ||
AK146865.1 | - | 15 - 16 | 1561 | 98 | ||
BC033050.1 | - | 2 - 3 | 273 | 98 | ||
BC040098.1 | - | 7 - 8 | 623 | 98 | ||
BC051500.1 | - | 3 - 4 | 430 | 98 | ||
BC058987.1 | - | 7 - 8 | 625 | 98 | ||
BC141193.1 | - | 15 - 16 | 1577 | 9055805 | 98 | |
BC141194.1 | - | 15 - 16 | 1577 | 9055806 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |