Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026873.2 | NP_081149.1 | 1 - 2 | 114 | 158 | |
GenBank mRNA | AK004502.1 | - | 1 - 2 | 124 | 158 | |
AK144396.1 | - | 1 - 2 | 126 | 158 | ||
BC016563.1 | - | 1 - 2 | 116 | 158 | ||
BC025110.1 | - | 1 - 2 | 114 | 3987809 | 158 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |