Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026873.2 | NP_081149.1 | 8 - 9 | 823 | 150 | |
GenBank mRNA | AK004502.1 | - | 8 - 9 | 832 | 150 | |
AK144396.1 | - | 8 - 9 | 834 | 150 | ||
BC016563.1 | - | 8 - 9 | 825 | 150 | ||
BC025110.1 | - | 8 - 9 | 823 | 3987809 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |