Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173757.3 | NP_776118.2 | 4 - 5 | 303 | 89 | |
GenBank mRNA | AK087686.1 | - | 4 - 5 | 299 | 89 | |
BC049149.1 | - | 4 - 5 | 230 | 6309524 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |