Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sclt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081411.1 | NP_001074880.1 | 2 - 3 | 585 | 75 | |
GenBank mRNA | AK011907.1 | - | 2 - 3 | 383 | 75 | |
AK014777.1 | - | 2 - 3 | 409 | 75 | ||
AK083894.1 | - | 2 - 3 | 568 | 75 | ||
BC043136.1 | - | 2 - 3 | 552 | 6812105 | 75 | |
BC060271.1 | - | 2 - 3 | 552 | 6812105 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |