Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028120.2 | NP_082396.1 | 10 - 11 | 1156 | 73 | |
GenBank mRNA | AK012067.1 | - | 10 - 11 | 1157 | 73 | |
AK029107.1 | - | 10 - 11 | 1187 | 73 | ||
BC003981.1 | - | 10 - 11 | 1211 | 3490246 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |