Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178912.3 | NP_849243.2 | 1 - 2 | 572 | 69 | |
GenBank mRNA | AK039944.1 | - | 1 - 2 | 576 | 69 | |
AK050306.1 | - | 1 - 2 | 573 | 69 | ||
AK053715.1 | - | 1 - 2 | 572 | 69 | ||
AK084697.1 | - | 1 - 2 | 576 | 69 | ||
AK086395.1 | - | 1 - 2 | 575 | 69 | ||
AK173211.1 | - | 1 - 2 | 551 | 69 | ||
BC150785.1 | - | 1 - 2 | 566 | 9087696 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |