Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178912.3 | NP_849243.2 | 4 - 5 | 976 | 105 | |
GenBank mRNA | AK039944.1 | - | 4 - 5 | 980 | 105 | |
AK050306.1 | - | 4 - 5 | 977 | 105 | ||
AK053715.1 | - | 4 - 5 | 976 | 105 | ||
AK084697.1 | - | 4 - 5 | 980 | 105 | ||
AK086395.1 | - | 4 - 5 | 979 | 105 | ||
AK173211.1 | - | 4 - 5 | 955 | 105 | ||
BC150785.1 | - | 4 - 5 | 970 | 9087696 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |