Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025522.5 | NP_079798.2 | 5 - 6 | 818 | 85 | |
GenBank mRNA | AK009385.1 | - | 5 - 6 | 797 | 85 | |
AK014100.1 | - | 5 - 6 | 773 | 85 | ||
BC016189.1 | - | 5 - 6 | 784 | 4482814 | 85 | |
CF586466.1 | - | 2 - 3 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |