Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mis18bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172578.2 | NP_766166.2 | 11 - 12 | 2963 | 104 | |
GenBank mRNA | AK032698.1 | - | 9 - 10 | 1995 | 104 | |
AK220223.1 | - | 10 - 11 | 2229 | 104 | ||
BC021832.1 | - | 6 - 7 | 1476 | 104 | ||
BC052175.1 | - | 11 - 12 | 2963 | 6394068 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |