Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153064.4 | NP_694704.1 | 3 - 4 | 416 | 69 | |
GenBank mRNA | AK078474.1 | - | 3 - 4 | 417 | 69 | |
AK146269.1 | - | 3 - 4 | 421 | 69 | ||
AK150431.1 | - | 3 - 4 | 412 | 69 | ||
AK151746.1 | - | 3 - 4 | 389 | 69 | ||
AK153364.1 | - | 3 - 4 | 389 | 69 | ||
AK165426.1 | - | 3 - 4 | 412 | 69 | ||
AK167016.1 | - | 3 - 4 | 414 | 69 | ||
AK168971.1 | - | 3 - 4 | 409 | 69 | ||
BC003898.1 | - | 3 - 4 | 415 | 69 | ||
BC016097.1 | - | 3 - 4 | 403 | 4910260 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |