Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146323.1 | NP_001139795.1 | 4 - 5 | 617 | 122 | |
NM_001146324.1 | NP_001139796.1 | 4 - 5 | 1013 | 122 | ||
NM_080634.4 | NP_542365.3 | 4 - 5 | 1013 | 122 | ||
GenBank mRNA | AF393747.1 | - | 4 - 5 | 1002 | 122 | |
AF393780.1 | - | 4 - 5 | 1005 | 122 | ||
AK034377.1 | - | 4 - 5 | 984 | 122 | ||
AK036663.1 | - | 4 - 5 | 1021 | 122 | ||
AK037829.1 | - | 4 - 5 | 512 | 122 | ||
AK049204.1 | - | 4 - 5 | 1006 | 122 | ||
AK079896.1 | - | 4 - 5 | 1005 | 122 | ||
AK149359.1 | - | 4 - 5 | 999 | 122 | ||
AK155497.1 | - | 4 - 5 | 980 | 122 | ||
AK169855.1 | - | 3 - 4 | 1085 | 122 | ||
AK170581.1 | - | 4 - 5 | 529 | 122 | ||
AK171997.1 | - | 4 - 5 | 605 | 122 | ||
BC058235.1 | - | 4 - 5 | 987 | 6307400 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |