Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mipol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164370.1 | NP_001157842.1 | 2 - 3 | 393 | 73 | |
GenBank mRNA | AK006974.1 | - | 302 | 73 | ||
AK020763.1 | - | 2 - 3 | 269 | 73 | ||
AK046422.1 | - | 2 - 3 | 391 | 73 | ||
AK050030.1 | - | 2 - 3 | 379 | 73 | ||
AK085510.1 | - | 2 - 3 | 388 | 73 | ||
AK163490.1 | - | 2 - 3 | 373 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |