Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pappa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085376.2 | NP_001078845.1 | 1 - 2 | 921 | 79 | |
GenBank mRNA | AK084006.1 | - | 1 - 2 | 432 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Protease Mus musculus Probe spans exons |