Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pappa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085376.2 | NP_001078845.1 | 2 - 3 | 1998 | 77 | |
GenBank mRNA | AK084006.1 | - | 2 - 3 | 1509 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Protease Mus musculus Probe spans exons |