Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013675.3 | NP_038703.3 | 16 - 17 | 4227 | 60 | |
GenBank mRNA | AK147299.1 | - | 16 - 17 | 4228 | 60 | |
AK147684.1 | - | 16 - 17 | 3820 | 60 | ||
AK147704.1 | - | 16 - 17 | 3820 | 60 | ||
BC129806.1 | - | 15 - 16 | 3560 | 40087684 | 60 | |
BC150783.1 | - | 15 - 16 | 3593 | 9087694 | 60 | |
M18641.1 | - | 1 - 2 | 374 | 60 | ||
S66283.1 | - | 16 - 17 | 3703 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |