Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nubpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029760.2 | NP_084036.2 | 3 - 4 | 325 | 81 | |
GenBank mRNA | AK010827.1 | - | 3 - 4 | 307 | 81 | |
AK141257.1 | - | 3 - 4 | 289 | 81 | ||
AK152377.1 | - | 3 - 4 | 326 | 81 | ||
BC062175.1 | - | 3 - 4 | 306 | 30291107 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |