Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC014689.1 | - | 2 - 3 | 417 | 69 | |
BC030904.1 | - | 1 - 2 | 257 | 69 | ||
BC034078.1 | - | 4 - 5 | 649 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |