Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Intu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175515.5 | NP_780724.3 | 1610 | 72 | ||
GenBank mRNA | AK035092.1 | - | 1591 | 72 | ||
AK122485.1 | - | 8 - 9 | 1150 | 72 | ||
BC125523.1 | - | 1557 | 40130038 | 72 | ||
BC132387.1 | - | 1557 | 40130664 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |